2,5-Dichloro-N-[4-(hexyloxy)benzyl]aniline

Supplier: Matrix Scientific

031793-500MG
101832-410EA 306.15 USD
101832-410
2,5-Dichloro-N-[4-(hexyloxy)benzyl]aniline
2,5-Dichloro-N-[4-(hexyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₁₉H₂₃Cl₂NO
MDL Number: MFCD10687822
CAS Number: 1040683-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR