2,5-Dichloro-N-[2-(2-chlorophenoxy)propyl]aniline

Supplier: Matrix Scientific

031792-500MG
101832-408EA 306.15 USD
101832-408
2,5-Dichloro-N-[2-(2-chlorophenoxy)propyl]aniline
2,5-Dichloro-N-[2-(2-chlorophenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₄Cl₃NO
MDL Number: MFCD10687821
CAS Number: 1040683-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR