2,5-Dichloro-N-(3-methylbenzyl)aniline

Supplier: Matrix Scientific

031788-500MG
101832-400EA 306.15 USD
101832-400
2,5-Dichloro-N-(3-methylbenzyl)aniline
2,5-Dichloro-N-(3-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₃Cl₂N
MDL Number: MFCD10687818
CAS Number: 1040084-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR