N-Butylaniline

Supplier: Matrix Scientific

031787-500MG
101832-398EA 306.15 USD
101832-398
N-Butylaniline
N-Butylaniline

Hazard: Irritant

Formula: (C₆H₅)NHCH₂CH₂CH₂CH₃
MW: 149.24 g/mol
MDL Number: MFCD00035798
CAS Number: 1126-78-9
UN: 2738
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR