N-(2-Isopropoxybenzyl)-2-methoxy-1-ethanamine

Supplier: Matrix Scientific

031784-500MG
101832-392EA 306.15 USD
101832-392
N-(2-Isopropoxybenzyl)-2-methoxy-1-ethanamine
N-(2-Isopropoxybenzyl)-2-methoxy-1-ethanamine

Hazard: Irritant

Formula: C₁₃H₂₁NO₂
MDL Number: MFCD10687815
CAS Number: 1040683-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR