N-(2-Methoxyethyl)-2-(4-methylphenoxy)-1-butanamine

Supplier: Matrix Scientific

031782-500MG
101832-388EA 306.15 USD
101832-388
N-(2-Methoxyethyl)-2-(4-methylphenoxy)-1-butanamine
N-(2-Methoxyethyl)-2-(4-methylphenoxy)-1-butanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO₂
MDL Number: MFCD10687813
CAS Number: 1040686-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR