2-Methoxy-N-[2-(1-naphthyl)ethyl]-1-ethanamine

Supplier: Matrix Scientific

031780-500MG
101832-384EA 306.15 USD
101832-384
2-Methoxy-N-[2-(1-naphthyl)ethyl]-1-ethanamine
2-Methoxy-N-[2-(1-naphthyl)ethyl]-1-ethanamine

Hazard: Irritant

Formula: C₁₅H₁₉NO
MDL Number: MFCD10687811
CAS Number: 1040686-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR