N-[2-(2,4-Dichlorophenoxy)propyl]-N-(2-methoxyethyl)amine

Supplier: Matrix Scientific

031778-500MG
101832-380EA 306.15 USD
101832-380
N-[2-(2,4-Dichlorophenoxy)propyl]-N-(2-methoxyethyl)amine
N-[2-(2,4-Dichlorophenoxy)propyl]-N-(2-methoxyethyl)amine

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO₂
MDL Number: MFCD10687809
CAS Number: 1040686-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR