N-(4-Chlorobenzyl)-2-methoxy-1-ethanamine

Supplier: Matrix Scientific

031775-500MG
101832-374EA 306.15 USD
101832-374
N-(4-Chlorobenzyl)-2-methoxy-1-ethanamine
N-(4-Chlorobenzyl)-2-methoxy-1-ethanamine

Hazard: Irritant

Formula: C₁₀H₁₄ClNO
MDL Number: MFCD05863673
CAS Number: 827328-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR