N-[2-(2-Methoxyethoxy)benzyl]-2,6-dimethylaniline

Supplier: Matrix Scientific

031772-500MG
101832-368EA 306.15 USD
101832-368
N-[2-(2-Methoxyethoxy)benzyl]-2,6-dimethylaniline
N-[2-(2-Methoxyethoxy)benzyl]-2,6-dimethylaniline

Hazard: Irritant

Formula: C₁₈H₂₃NO₂
MDL Number: MFCD10687806
CAS Number: 1040686-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR