N-(2-Fluorobenzyl)-2,6-dimethylaniline

Supplier: Matrix Scientific

031771-500MG
101832-366EA 306.15 USD
101832-366
N-(2-Fluorobenzyl)-2,6-dimethylaniline
N-(2-Fluorobenzyl)-2,6-dimethylaniline

Hazard: Irritant

Formula: C₁₅H₁₆FN
MDL Number: MFCD10687805
CAS Number: 1040686-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR