N-(Cyclopropylmethyl)-2,6-dimethylaniline

Supplier: Matrix Scientific

031767-500MG
101832-358EA 306.15 USD
101832-358
N-(Cyclopropylmethyl)-2,6-dimethylaniline
N-(Cyclopropylmethyl)-2,6-dimethylaniline

Hazard: Irritant

Formula: C₁₂H₁₇N
MDL Number: MFCD10687802
CAS Number: 1040686-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR