N-[2-(3,5-Dimethylphenoxy)ethyl]-2,6-dimethylaniline

Supplier: Matrix Scientific

031763-500MG
101832-350EA 306.15 USD
101832-350
N-[2-(3,5-Dimethylphenoxy)ethyl]-2,6-dimethylaniline
N-[2-(3,5-Dimethylphenoxy)ethyl]-2,6-dimethylaniline

Hazard: Irritant

Formula: C₁₈H₂₃NO
MDL Number: MFCD10687798
CAS Number: 1040686-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR