N-[2-(4-Chloro-3-methylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031760-500MG
101832-344EA 306.15 USD
101832-344
N-[2-(4-Chloro-3-methylphenoxy)ethyl]aniline
N-[2-(4-Chloro-3-methylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₆ClNO
MDL Number: MFCD10687795
CAS Number: 1040686-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR