2-Fluoro-N-[4-(2-phenoxyethoxy)benzyl]aniline

Supplier: Matrix Scientific

031759-500MG
101832-342EA 306.15 USD
101832-342
2-Fluoro-N-[4-(2-phenoxyethoxy)benzyl]aniline
2-Fluoro-N-[4-(2-phenoxyethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₁H₂₀FNO₂
MDL Number: MFCD10687794
CAS Number: 1040685-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR