N-[2-(4-Chloro-2-methylphenoxy)ethyl]-2-fluoroaniline

Supplier: Matrix Scientific

031755-500MG
101832-334EA 306.15 USD
101832-334
N-[2-(4-Chloro-2-methylphenoxy)ethyl]-2-fluoroaniline
N-[2-(4-Chloro-2-methylphenoxy)ethyl]-2-fluoroaniline

Hazard: Irritant

Formula: C₁₅H₁₅ClFNO
MDL Number: MFCD10687790
CAS Number: 1040685-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR