N-[2-(2,4-Dichlorophenoxy)ethyl]-2-fluoroaniline

Supplier: Matrix Scientific

031754-500MG
101832-332EA 592.31 USD
101832-332
N-[2-(2,4-Dichlorophenoxy)ethyl]-2-fluoroaniline
N-[2-(2,4-Dichlorophenoxy)ethyl]-2-fluoroaniline

Hazard: Irritant

Formula: C₁₄H₁₂Cl₂FNO
MDL Number: MFCD10687789
CAS Number: 1040685-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR