3-Chloro-N-[2-(2,5-dimethylphenoxy)propyl]-4-fluoroaniline

Supplier: Matrix Scientific

031749-500MG
101832-322EA 306.15 USD
101832-322
3-Chloro-N-[2-(2,5-dimethylphenoxy)propyl]-4-fluoroaniline
3-Chloro-N-[2-(2,5-dimethylphenoxy)propyl]-4-fluoroaniline

Hazard: Irritant

Formula: C₁₇H₁₉ClFNO
MDL Number: MFCD10687784
CAS Number: 1040685-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR