N-(3-Chloro-4-fluorophenyl)-N-[2-(2-methyl-phenoxy)butyl]amine

Supplier: Matrix Scientific

031748-500MG
101832-320EA 306.15 USD
101832-320
N-(3-Chloro-4-fluorophenyl)-N-[2-(2-methyl-phenoxy)butyl]amine
N-(3-Chloro-4-fluorophenyl)-N-[2-(2-methyl-phenoxy)butyl]amine

Hazard: Irritant

Formula: C₁₇H₁₉ClFNO
MDL Number: MFCD10687783
CAS Number: 1040685-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR