3-Chloro-4-fluoro-N-(2-methoxybenzyl)aniline

Supplier: Matrix Scientific

031747-500MG
101832-318EA 306.15 USD
101832-318
3-Chloro-4-fluoro-N-(2-methoxybenzyl)aniline
3-Chloro-4-fluoro-N-(2-methoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₃ClFNO
MDL Number: MFCD01826283
CAS Number: 416869-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR