N-(2-Isopropoxybenzyl)-1-propanamine

Supplier: Matrix Scientific

031741-500MG
101832-306EA 306.15 USD
101832-306
N-(2-Isopropoxybenzyl)-1-propanamine
N-(2-Isopropoxybenzyl)-1-propanamine

Hazard: Irritant

Formula: C₁₃H₂₁NO
MDL Number: MFCD10687778
CAS Number: 1040685-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR