N-(4-Isobutoxybenzyl)-1-propanamine

Supplier: Matrix Scientific

031740-500MG
101832-304EA 306.15 USD
101832-304
N-(4-Isobutoxybenzyl)-1-propanamine
N-(4-Isobutoxybenzyl)-1-propanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10687777
CAS Number: 1040685-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR