2-(2-Methylphenoxy)-N-propyl-1-butanamine

Supplier: Matrix Scientific

031739-500MG
101832-302EA 306.15 USD
101832-302
2-(2-Methylphenoxy)-N-propyl-1-butanamine
2-(2-Methylphenoxy)-N-propyl-1-butanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10687776
CAS Number: 1040685-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR