N-[2-(4-Methoxyphenoxy)ethyl]-1-propanamine

Supplier: Matrix Scientific

031738-500MG
101832-300EA 306.15 USD
101832-300
N-[2-(4-Methoxyphenoxy)ethyl]-1-propanamine
N-[2-(4-Methoxyphenoxy)ethyl]-1-propanamine

Hazard: Irritant

Formula: C₁₂H₁₉NO₂
MDL Number: MFCD10687775
CAS Number: 1040314-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR