N-(4-Chlorobenzyl)-1-propanamine

Supplier: Matrix Scientific

031737-500MG
101832-298EA 306.15 USD
101832-298
N-(4-Chlorobenzyl)-1-propanamine
N-(4-Chlorobenzyl)-1-propanamine

Hazard: Irritant

Formula: C₁₀H₁₄ClN
MDL Number: MFCD04568716
CAS Number: 55245-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR