N-[2-(2-Isopropylphenoxy)ethyl]-1-propanamine

Supplier: Matrix Scientific

031736-500MG
101832-296EA 306.15 USD
101832-296
N-[2-(2-Isopropylphenoxy)ethyl]-1-propanamine
N-[2-(2-Isopropylphenoxy)ethyl]-1-propanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10687774
CAS Number: 1038376-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR