2-Furyl-N-[3-(phenethyloxy)benzyl]methanamine

Supplier: Matrix Scientific

031735-500MG
101832-294EA 306.15 USD
101832-294
2-Furyl-N-[3-(phenethyloxy)benzyl]methanamine
2-Furyl-N-[3-(phenethyloxy)benzyl]methanamine

Hazard: Irritant

Formula: C₂₀H₂₁NO₂
MDL Number: MFCD10687773
CAS Number: 1040684-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR