2-Furyl-N-(4-isobutoxybenzyl)methanamine

Supplier: Matrix Scientific

031734-500MG
101832-292EA 306.15 USD
101832-292
2-Furyl-N-(4-isobutoxybenzyl)methanamine
2-Furyl-N-(4-isobutoxybenzyl)methanamine

Hazard: Irritant

Formula: C₁₆H₂₁NO₂
MDL Number: MFCD10687772
CAS Number: 898507-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR