N-[2-(4-Ethylphenoxy)propyl]-3-methylaniline
Supplier: Matrix Scientific
031733-500MG
101832-290EA
306.15
USD
101832-290
N-[2-(4-Ethylphenoxy)propyl]-3-methylaniline
N-[2-(4-Ethylphenoxy)propyl]-3-methylaniline
Hazard: Irritant
Formula:
C₁₈H₂₃NO |
MDL Number:
MFCD10687771 CAS Number: 1040684-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...