N-[2-(2,5-Dimethylphenoxy)propyl]-N-(2-furylmethyl)amine

Supplier: Matrix Scientific

031732-500MG
101832-288EA 306.15 USD
101832-288
N-[2-(2,5-Dimethylphenoxy)propyl]-N-(2-furylmethyl)amine
N-[2-(2,5-Dimethylphenoxy)propyl]-N-(2-furylmethyl)amine

Hazard: Irritant

Formula: C₁₆H₂₁NO₂
MDL Number: MFCD10687770
CAS Number: 1040684-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR