2-(4-Ethylphenoxy)-N-(2-furylmethyl)-1-ethanamine hydrochloride

Supplier: Matrix Scientific

031729-500MG
101832-282EA 306.15 USD
101832-282
2-(4-Ethylphenoxy)-N-(2-furylmethyl)-1-ethanamine hydrochloride
2-(4-Ethylphenoxy)-N-(2-furylmethyl)-1-ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₀ClNO₂
MDL Number: MFCD20921470
CAS Number: 1357252-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR