N-(2-Furylmethyl)-2-phenoxy-1-ethanamine

Supplier: Matrix Scientific

031727-500MG
101832-280EA 306.15 USD
101832-280
N-(2-Furylmethyl)-2-phenoxy-1-ethanamine
N-(2-Furylmethyl)-2-phenoxy-1-ethanamine

Hazard: Irritant

Formula: C₁₃H₁₅NO₂
MDL Number: MFCD10687766
CAS Number: 82395-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR