N-(2-Chlorobenzyl)-2-ethoxyaniline

Supplier: Matrix Scientific

031720-500MG
101832-266EA 306.15 USD
101832-266
N-(2-Chlorobenzyl)-2-ethoxyaniline
N-(2-Chlorobenzyl)-2-ethoxyaniline

Hazard: Irritant

Formula: C₁₅H₁₆ClNO
MDL Number: MFCD10687760
CAS Number: 1019498-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR