N-[3-(Isopentyloxy)benzyl]-2-methoxyaniline

Supplier: Matrix Scientific

031719-500MG
101832-264EA 306.15 USD
101832-264
N-[3-(Isopentyloxy)benzyl]-2-methoxyaniline
N-[3-(Isopentyloxy)benzyl]-2-methoxyaniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687759
CAS Number: 1040684-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR