N-[4-(tert-Butyl)benzyl]-2-methoxyaniline

Supplier: Matrix Scientific

031715-500MG
101832-256EA 306.15 USD
101832-256
N-[4-(tert-Butyl)benzyl]-2-methoxyaniline
N-[4-(tert-Butyl)benzyl]-2-methoxyaniline

Hazard: Irritant

Formula: C₁₈H₂₃NO
MDL Number: MFCD10687755
CAS Number: 1040683-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR