2-Methoxy-N-[2-(2-methylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031713-500MG
101832-252EA 306.15 USD
101832-252
2-Methoxy-N-[2-(2-methylphenoxy)ethyl]aniline
2-Methoxy-N-[2-(2-methylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₆H₁₉NO₂
MDL Number: MFCD10687754
CAS Number: 1040683-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR