Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(3-Isopropoxybenzyl)-2-propanamine

Supplier: Matrix Scientific


031709-500MG
101832-244EA 260.23 USD InStock
101832-244
N-(3-Isopropoxybenzyl)-2-propanamine
N-(3-Isopropoxybenzyl)-2-propanamine
CAS: 1040683-60-0
MDL Number: MFCD10687750
Molecular Formula: C13H21NO
Molecular Weight: 207,32
Hazard: Irritant


Formula: C₁₃H₂₁NO
MW: 207,32 g/mol
MDL Number: MFCD10687750
CAS Number: 1040683-60-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty