N-(3-Isobutoxybenzyl)-2-propanamine

Supplier: Matrix Scientific

031708-500MG
101832-242EA 306.15 USD
101832-242
N-(3-Isobutoxybenzyl)-2-propanamine
N-(3-Isobutoxybenzyl)-2-propanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10687749
CAS Number: 1040683-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR