N-[2-(Pentyloxy)benzyl]-2-propanamine

Supplier: Matrix Scientific

031706-500MG
101832-238EA 306.15 USD
101832-238
N-[2-(Pentyloxy)benzyl]-2-propanamine
N-[2-(Pentyloxy)benzyl]-2-propanamine

Hazard: Irritant

Formula: C₁₅H₂₅NO
MDL Number: MFCD10687747
CAS Number: 1040683-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR