N-(2-Butoxybenzyl)-2-propanamine

Supplier: Matrix Scientific

031705-500MG
101832-236EA 306.15 USD
101832-236
N-(2-Butoxybenzyl)-2-propanamine
N-(2-Butoxybenzyl)-2-propanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD10687746
CAS Number: 1040683-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR