N-(2-Methoxybenzyl)-2-propanamine

Supplier: Matrix Scientific

Synonyms: N-(2-Methoxybenzyl)propan-2-amine

031702-500MG
101832-230EA 306.15 USD
101832-230
N-(2-Methoxybenzyl)-2-propanamine
N-(2-Methoxybenzyl)-2-propanamine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MW: 179.26 g/mol
MDL Number: MFCD08691655
CAS Number: 756474-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR