N-[2-(2,5-Dimethylphenoxy)ethyl]-2-propanamine

Supplier: Matrix Scientific

031701-500MG
101832-228EA 306.15 USD
101832-228
N-[2-(2,5-Dimethylphenoxy)ethyl]-2-propanamine
N-[2-(2,5-Dimethylphenoxy)ethyl]-2-propanamine

Hazard: Irritant

Formula: C₁₃H₂₁NO
MDL Number: MFCD10687744
CAS Number: 1040017-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR