N-(4-Methoxyphenethyl)-2-propanamine

Supplier: Matrix Scientific

031700-500MG
101832-226EA 306.15 USD
101832-226
N-(4-Methoxyphenethyl)-2-propanamine
N-(4-Methoxyphenethyl)-2-propanamine

Hazard: Irritant

Formula: C₁₂H₁₉NO
MDL Number: MFCD05863687
CAS Number: 884497-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR