N-(2-Phenoxyethyl)-2-propanamine

Supplier: Matrix Scientific

031698-500MG
101832-222EA 306.15 USD
101832-222
N-(2-Phenoxyethyl)-2-propanamine
N-(2-Phenoxyethyl)-2-propanamine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD08691703
CAS Number: 55247-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR