N-[2-(4-Ethylphenoxy)ethyl]-3-methylaniline

Supplier: Matrix Scientific

031696-500MG
101832-218EA 306.15 USD
101832-218
N-[2-(4-Ethylphenoxy)ethyl]-3-methylaniline
N-[2-(4-Ethylphenoxy)ethyl]-3-methylaniline

Hazard: Irritant

Formula: C₁₇H₂₁NO
MDL Number: MFCD10687742
CAS Number: 1040683-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR