N-[2-(2-Methoxyphenoxy)propyl]-2,3-dimethylaniline

Supplier: Matrix Scientific

031692-500MG
101832-210EA 306.15 USD
101832-210
N-[2-(2-Methoxyphenoxy)propyl]-2,3-dimethylaniline
N-[2-(2-Methoxyphenoxy)propyl]-2,3-dimethylaniline

Hazard: Irritant

Formula: C₁₈H₂₃NO₂
MDL Number: MFCD10687738
CAS Number: 1040682-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR