N-(3-Chlorobenzyl)-2,3-dimethylaniline

Supplier: Matrix Scientific

031691-500MG
101832-208EA 306.15 USD
101832-208
N-(3-Chlorobenzyl)-2,3-dimethylaniline
N-(3-Chlorobenzyl)-2,3-dimethylaniline

Hazard: Irritant

Formula: C₁₅H₁₆ClN
MDL Number: MFCD06408364
CAS Number: 1024246-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR