N-(2-Methoxybenzyl)-2,3-dimethylaniline

Supplier: Matrix Scientific

031689-500MG
101832-204EA 306.15 USD
101832-204
N-(2-Methoxybenzyl)-2,3-dimethylaniline
N-(2-Methoxybenzyl)-2,3-dimethylaniline

Hazard: Irritant

Formula: C₁₆H₁₉NO
MDL Number: MFCD10687736
CAS Number: 1020939-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR