N-[2-(4-Chloro-3-methylphenoxy)ethyl]-2,3-dimethylaniline

Supplier: Matrix Scientific

031686-500MG
101832-198EA 306.15 USD
101832-198
N-[2-(4-Chloro-3-methylphenoxy)ethyl]-2,3-dimethylaniline
N-[2-(4-Chloro-3-methylphenoxy)ethyl]-2,3-dimethylaniline

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD10687733
CAS Number: 1040682-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR