N-([1,1'-Biphenyl]-4-ylmethyl)-2,3-dimethylaniline

Supplier: Matrix Scientific

031685-500MG
101832-196EA 306.15 USD
101832-196
N-([1,1'-Biphenyl]-4-ylmethyl)-2,3-dimethylaniline
N-([1,1'-Biphenyl]-4-ylmethyl)-2,3-dimethylaniline

Hazard: Irritant

Formula: C₂₁H₂₁N
MDL Number: MFCD10687732
CAS Number: 1040682-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR